MilliporeSigma
  • Comparison of three different plasma homocysteine assays with gas chromatography-mass spectrometry.
Comparison of three different plasma homocysteine assays with gas chromatography-mass spectrometry.
Clinical chemistry (1999-05-01)
J B Ubbink, R Delport, R Riezler, W J Vermaak
ABSTRACT
Various methods are available to measure plasma total homocyst(e)ine (tHcy) concentrations, but whether plasma tHcy assays may be used interchangeably is not known. Results from three different methods [HPLC with fluorescence detection, enzyme immunoassay (EIA), and fluorescence polarization immunoassay (FPIA)] to determine fasting (n = 163) and post-methionine load (n = 80) plasma tHcy concentrations were compared with those obtained by gas chromatography-mass spectrometry (GC-MS). Difference plots on non-transformed and log-transformed data were used to assess the agreement between HPLC and GC-MS, EIA and GC-MS, and FPIA and GC-MS. The closest agreement between methods was observed between GC-MS and FPIA for fasting tHcy concentrations, with 95% of the FPIA values between 19% above and 24% below the corresponding GC-MS results. Post-methionine load tHcy concentrations measured by EIA showed the least agreement with GC-MS, with 95% of values measured by EIA ranging between 52% above and 16% below the GC-MS values. With respect to GC-MS, the above-mentioned methods showed a negative bias for fasting tHcy concentrations, but a positive bias for both immunoassays for post-methionine load tHcy concentrations. The agreement among methods is insufficient to allow them to be used interchangeably. The intermethod differences emphasize the need for standardization of plasma tHcy assays.
Sigma-Aldrich
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, ≥98%
Supelco
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, for HPLC derivatization, LiChropur, ≥98.5% (HPLC)