MilliporeSigma
  • Anticonvulsant effects of eliprodil alone or combined with the glycineB receptor antagonist L-701,324 or the competitive NMDA antagonist CGP 40116 in the amygdala kindling model in rats.
Anticonvulsant effects of eliprodil alone or combined with the glycineB receptor antagonist L-701,324 or the competitive NMDA antagonist CGP 40116 in the amygdala kindling model in rats.
Neuropharmacology (1999-04-28)
P Wlaź, U Ebert, W Löscher
ABSTRACT
The discovery that glutamate's activity at the N-methyl-D-aspartate (NMDA) receptor is positively modulated by glycine and polyamines has led to a new pharmacological strategy that NMDA receptor-mediated events could be antagonized indirectly at the strychnine-insensitive glycine co-agonist site (glycine(B) receptor) and the polyamine modulatory site. Recently we demonstrated that ifenprodil and L-701,324 (7-chloro-4-hydroxy-3(3-phenoxy)phenyl-2(H)quinoline), polyamine and glycine, receptor antagonists, respectively, at subeffective doses markedly increased after-discharge threshold (ADT) when applied together in amygdala-kindled rats. Because ifenprodil and its derivative, eliprodil, exhibit different affinities for NMDA receptors composed of different subunits, our current question was whether a combination of eliprodil and the glycine, receptor antagonist, L-701,324, would produce a super-additive anticonvulsant action. In addition, we examined the combined treatment of eliprodil with a competitive NMDA receptor antagonist CGP 40116 (D-(E)-2-amino-4-methyl-5-phosphono-3-pentenoic acid) in the kindling model. Eliprodil alone (10-40 mg/kg) had no consistent ADT-increasing activity. When eliprodil was combined with an ineffective dose of L-701,324 (2.5 mg/kg), a significant rise in ADT was observed. Likewise, other measures of seizure activity such as severity and duration were modestly but significantly reduced. With respect to behavioral impairments, no signs of synergistic interaction were observed after the drug combinations. On the other hand, no anticonvulsant effects were found when CGP 40116 was administered alone at doses of 1.25-5 mg/kg or CGP 40116 1.25 mg/kg combined with eliprodil 10 mg/kg. These data suggest that combination therapy with antagonists at the polyamine and glycine sites might potentially treat therapy-resistant complex partial seizures.
Sigma-Aldrich
Eliprodil, ≥98% (HPLC), powder