MilliporeSigma
  • Protective effects of vasoactive intestinal peptide against delayed glutamate neurotoxicity in cultured retina.
Protective effects of vasoactive intestinal peptide against delayed glutamate neurotoxicity in cultured retina.
Brain research (1998-10-31)
K Shoge, H K Mishima, T Saitoh, K Ishihara, Y Tamura, H Shiomi, M Sasa
ABSTRACT
The effects of vasoactive intestinal peptide (VIP) on glutamate-induced delayed death were examined using the primary cultures of rat retinal neurons. Effects of VIP on glutamate-induced neurotoxicity were evaluated by double staining with fluorescein diacetate and propidium iodide. Glutamate (1 mM) was applied to the culture for 10 min in the presence and absence of VIP, and visible cells enumerated 24 h after culture in normal medium. Effects of VIP on increase in the intracellular Ca2+ concentration and currents induced by glutamate in retinal neurons were investigated using the Ca2+ image analyzing system with fura-2 and whole-cell patch-clamp recording, respectively. The cAMP contents in retinal cultures were measured by radioimmunoassay. VIP (10 nM-1 microM) dose-dependently protected against glutamate-induced neurotoxicity in cultured retinal neurons. Protection by VIP (100 nM) against glutamate (1 mM)-induced neurotoxicity was antagonized by VIP6-28 (1 microM), a VIP antagonist, and H-89 (100 nM and 1 microM), a protein kinase A inhibitor. However, VIP had no effect on glutamate-induced inward currents nor glutamate-induced increase in the intracellular Ca2+ concentration. A 10-min exposure of VIP (100 nM) with glutamate (1 mM) resulted in an increase in the cAMP level to 446+/-58 from 22+/-1 pmol/mg protein. These findings suggest that VIP protects against the glutamate-induced neurotoxicity in retinal cultures by elevating the cAMP level via VIP receptors and thereby activating protein kinase A.
Sigma-Aldrich
Vasoactive Intestinal Peptide Fragment 6-28 human, porcine, rat, ≥97% (HPLC)