MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Characterization of recombinant soluble human transforming growth factor-beta receptor type II (rhTGF-beta sRII).
Characterization of recombinant soluble human transforming growth factor-beta receptor type II (rhTGF-beta sRII).
Cytokine (1995-07-01)
M L Tsang, L Zhou, B L Zheng, J Wenker, G Fransen, J Humphrey, J M Smith, M O'Connor-McCourt, R Lucas, J A Weatherbee
ABSTRACT
Recombinant human transforming growth factor soluble receptor Type II (rhTGF-beta sRII) corresponding to the 159 amino acid extracellular domain of hTGF-beta RII has been expressed in insect cells using the baculovirus expression system or in a mouse myeloma cell line. N-terminal sequence analysis of the purified protein revealed the removal of the 23 amino acid signal peptide. In SDS-PAGE, the rhTGF-beta sRII resolves into multiple bands due to N-linked glycosylation. Recombinant hTGF-beta sRII is a TGF-beta antagonist and will inhibit the biological activities of TGF-beta 1, TGF-beta 3, and TGF-beta 5 on TGF-beta-responsive cell lines, such as murine HT-2 or human TF-1 with an ED50 of approximately 0.3 micrograms/mL. However, hTGF-beta sRII does not inhibit TGF-beta 2 bioactivities in these cell lines, suggesting that hTGF-beta RII has low affinity for TGF-beta 2. Polyclonal antibodies to hTGF-beta sRII have been produced in goats and purified on Protein-G affinity columns. This antibody can inhibit TGF-beta 1,2,3,5-dependent bioactivities on human cell lines such as TF-1. Additionally, this antibody has species cross-reactivity and will also inhibit TGF-beta-dependent bioactivities on murine cells.(ABSTRACT TRUNCATED AT 250 WORDS)