MilliporeSigma
  • Demonstration of peptide:N-glycosidase F activity in endo-beta-N-acetylglucosaminidase F preparations.
Demonstration of peptide:N-glycosidase F activity in endo-beta-N-acetylglucosaminidase F preparations.
The Journal of biological chemistry (1984-09-10)
T H Plummer, J H Elder, S Alexander, A W Phelan, A L Tarentino
ABSTRACT
Endo-beta-N-acetylglucosaminidase F preparations from Flavobacterium meningosepticum have been found to contain peptide:N-glycosidase activity. Only the second activity, designated as peptide:N-glycosidase F, readily cleaves the beta-aspartylglycosylamine linkage of a fetuin triantennary complex glycopeptide, as shown by the isolation of the corresponding carbohydrate-free peptide containing aspartic acid and of an intact oligosaccharide with a di-N-acetylchitobiosyl moiety at the reducing end. Both activities in the mixture will hydrolyze a high mannose octaglycopeptide from ovalbumin, with the type of product formed being influenced by pH. At pH 4.0, only the endo-beta-N-acetylglucosaminidase F activity is functional, releasing octapeptide-GlcNAc and oligosaccharide-GlcNAc. At pH 9.3, the predominant cleavage is by peptide:N-glycosidase F at the glycosylamine bond, releasing octapeptide and oligosaccharide-GlcNAc-GlcNAc. This latter oligosaccharide is then hydrolyzed by endo-beta-N-acetylglucosaminidase F to oligosaccharide-GlcNAc plus GlcNAc.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli