MilliporeSigma
  • Effects of gonadotropin-releasing hormone agonist on the incidence of apoptosis in porcine and human granulosa cells.
Effects of gonadotropin-releasing hormone agonist on the incidence of apoptosis in porcine and human granulosa cells.
Gynecologic and obstetric investigation (2000-01-12)
S Zhao, H Saito, X Wang, T Saito, T Kaneko, M Hiroi
ABSTRACT
Gonadotropin-releasing hormone (GnRH) has been found to be expressed in ovaries of various species and to modulate cell differentiation in ovarian cells. GnRH agonists (GnRHa) are widely used in the stimulation protocols of assisted reproduction. In the present study, the direct effects of a GnRHa on the incidence of apoptosis and sterodogenesis in porcine in human granulosa cells were investigated. Cells were obtained from 6-month-old swine and in vitro fertilization patients, and cultured in minimum essential medium (MEM) supplemented with 5% fetal bovine serum for 24 h. The GnRHa (burserelin acetate) was added to the MEM at various concentrations (0, 5, 50, 500 pg/ml, and 5 ng/ml). The nuclei of granulosa cells were examined by fluorescence microscopy after Hoechst 33258 staining, and the incidence of apoptotic cells was tabulated. The GnRHa directly increased the incidence of apoptosis in both types of granulosa cells. The concentration of GnRHa in clinical use is much higher than the concentration which produced apoptosis in granulosa cells here. The clinical dosage of GnRHa should be reconsidered with regard to its apoptosis-inducing effect.
Sigma-Aldrich
Buserelin acetate salt, ≥90% (HPLC)