MilliporeSigma
GABA transporter heterogeneity: pharmacology and cellular localization.
Neurochemistry international (1996-10-01)
L A Borden
ABSTRACT
gamma-Aminobutyric acid (GABA) is the major inhibitory neurotransmitter in the mammalian brain. GABA is cleared from the synaptic cleft by specific, high-affinity, sodium- and chloride-dependent transporters, which are thought to be located on presynaptic terminals and surrounding glial cells. While early studies suggested a distinction between neuronal and glial GABA transport, molecular cloning has revealed the existence of genes for four distinct GABA transporters (termed GAT-1, GAT-2, GAT-3 and BGT-1), thus revealing a greater heterogeneity than previously suspected. Heterologous expression has allowed a detailed characterization of their pharmacological properties, and has revealed that GAT-1 is the site of action of the anticonvulsant drug, Tiagabine. In-situ hybridization and immunocytochemistry demonstrate that each transporter has a unique regional distribution in the brain; in conjunction with experiments utilizing cell cultures, the neuronal vs glial localization of the various transporters is being elucidated. Future studies will be directed at determining the role of each transporter in the regulation of GABAergic transmission, and in the design of additional subtype-specific inhibitors, which may serve as novel therapeutic agents for the treatment of neuropsychiatric disorders.
Sigma-Aldrich
(S)-SNAP-5114, ≥98% (HPLC), solid