MilliporeSigma
  • G9a mediates Sharp-1-dependent inhibition of skeletal muscle differentiation.
G9a mediates Sharp-1-dependent inhibition of skeletal muscle differentiation.
Molecular biology of the cell (2012-10-23)
Belinda Mei Tze Ling, Suma Gopinadhan, Wai Kay Kok, Shilpa Rani Shankar, Pooja Gopal, Narendra Bharathy, Yaju Wang, Reshma Taneja
ABSTRACT
Sharp-1, a basic helix-loop-helix transcription factor, is a potent repressor of skeletal muscle differentiation and is dysregulated in muscle pathologies. However, the mechanisms by which it inhibits myogenesis are not fully understood. Here we show that G9a, a lysine methyltransferase, is involved in Sharp-1-mediated inhibition of muscle differentiation. We demonstrate that G9a directly interacts with Sharp-1 and enhances its ability to transcriptionally repress the myogenin promoter. Concomitant with a differentiation block, G9a-dependent histone H3 lysine 9 dimethylation (H3K9me2) and MyoD methylation are apparent upon Sharp-1 overexpression in muscle cells. RNA interference-mediated reduction of G9a or pharmacological inhibition of its activity erases these repressive marks and rescues the differentiation defect imposed by Sharp-1. Our findings provide new insights into Sharp-1-dependent regulation of myogenesis and identify epigenetic mechanisms that could be targeted in myopathies characterized by elevated Sharp-1 levels.
Sigma-Aldrich
UNC0638 hydrate, ≥98% (HPLC)