MilliporeSigma
  • Reactive oxygen species-mediated endoplasmic reticulum stress and mitochondrial dysfunction contribute to cirsimaritin-induced apoptosis in human gallbladder carcinoma GBC-SD cells.
Reactive oxygen species-mediated endoplasmic reticulum stress and mitochondrial dysfunction contribute to cirsimaritin-induced apoptosis in human gallbladder carcinoma GBC-SD cells.
Cancer letters (2010-04-03)
Zhiwei Quan, Jun Gu, Ping Dong, Jianhua Lu, Xiangsong Wu, Wenguang Wu, Xiaozhou Fei, Songgang Li, Yong Wang, Jianwei Wang, Yingbin Liu
ABSTRACT
In this study, the anticancer effect of cirsimaritin, a natural flavonoid, against human gallbladder carcinoma cell line GBC-SD and the underlying mechanisms were investigated. Cirsimaritin inhibited the growth of tumor cells and induced mitochondrial apoptosis in GBC-SD cells. In addition, cirsimaritin triggered endoplasmic reticulum (ER) stress and down-regulated the phosphorylation of Akt, while knock-down of CHOP dramatically abrogated the inactivation of Akt and reversed the pro-apoptotic effect of cirsimaritin. Furthermore, cirsimaritin provoked the generation of reactive oxygen species in GBC-SD cells, while the antioxidant N-acetyl cysteine almost completely blocked the activation of ER stress and apoptosis, suggesting cirsimaritin-induced reactive oxygen species is an early event that triggers ER stress mitochondrial apoptotic pathways in GBC-SD cells.
Sigma-Aldrich
Cirsimaritin, ≥90% (LC/MS-ELSD)