MilliporeSigma
  • Highly enantioselective synthesis of gamma-nitro heteroaromatic ketones in a doubly stereocontrolled manner catalyzed by bifunctional thiourea catalysts based on dehydroabietic amine: a doubly stereocontrolled approach to pyrrolidine carboxylic acids.
Highly enantioselective synthesis of gamma-nitro heteroaromatic ketones in a doubly stereocontrolled manner catalyzed by bifunctional thiourea catalysts based on dehydroabietic amine: a doubly stereocontrolled approach to pyrrolidine carboxylic acids.
Organic letters (2008-12-11)
Xianxing Jiang, Yifu Zhang, Albert S C Chan, Rui Wang
ABSTRACT
A new class of dehydroabietic amine-substituted primary amine-thiourea bifunctional catalysts were designed and synthesized. The doubly stereocontrolled organocatalytic conjugate addition of a variety of heterocycles-bearing ketones to nitroalkenes was investigated for the first time, affording (S)- or (R)-gamma-nitro heteroaromatic ketones with excellent enantioselectivities (up to ee >99%). Furthermore, the nearly optically pure gamma-nitro heteroaromatic ketones can be readily transformed into chiral pyrrolidine carboxylic acids.
Sigma-Aldrich
Dehydroabietic acid, ≥90% (LC/MS-ELSD)