MilliporeSigma
  • Effect of spores of saprophytic fungi on phytoalexin accumulation in seeds of frog-eye leaf spot and stem canker-resistant and -susceptible soybean (Glycine max L.) cultivars.
Effect of spores of saprophytic fungi on phytoalexin accumulation in seeds of frog-eye leaf spot and stem canker-resistant and -susceptible soybean (Glycine max L.) cultivars.
Journal of agricultural and food chemistry (2000-08-24)
W S Garcez, D Martins, F R Garcez, M R Marques, A A Pereira, L A Oliveira, J N Rondon, A D Peruca
ABSTRACT
Two saprophytic fungi (Mucor ramosissimus and Rhizopus sp.) were tested for their ability to induce phytoalexin production by seeds of frog-eye leaf spot and stem canker-resistant and -susceptible soybean (Glycine max L.) cultivars. Only M. ramosissimus was shown to elicit a response and qualitative differences in phytoalexin accumulation were found between the susceptible and resistant cultivars. Glyceollins I, II, and III and glycinol were isolated from the susceptible cultivar, whereas Glyceollins I, II, and III, glycinol, glyceocarpin, genistein, isoformononetin, and N-acetyltyramine accumulated in the resistant cultivar in response to the same fungal elicitor. Genistein was found to be an inducibly formed isoflavonoid instead of a constitutive metabolite in the resistant cultivar, whereas N-acetyltyramine is described for the first time as a soybean phytoalexin. All the compounds, except genistein, showed fungitoxic activity against Cladosporium sphaerospermum. Spectral data of the pterocarpan phytoalexins, genistein, and N-acetyltyramine are also given in this work.
Sigma-Aldrich
N-Acetyltyramine, ≥95% (LC/MS-ELSD)