MilliporeSigma
Secretion of thioredoxin after in vitro activation of human B cells.
Lymphokine and cytokine research (1992-10-01)
M L Ericson, J Hörling, V Wendel-Hansen, A Holmgren, A Rosén
ABSTRACT
The redox-active enzyme thioredoxin (Trx) is secreted by various virus-transformed cell lines of B- and T-cell origin and has been considered to play an autoregulatory role as a cofactor during cellular growth processes. We show in this paper that exposure of B lymphocytes from normal, healthy donors and B cells from B-type chronic lymphocytic leukemia (B-CLL) to Staphylococcus aureus Cowan I (SAC) induced expression of Trx mRNA. By combining SAC, or the phorbol ester TPA, with IL-2 and the conditioned medium of a T-cell hybridoma (BSF-MP6), we could strongly enhance the Trx expression. After [35S]methionine labeling of stimulated B-CLL cells in vitro, Trx was immunoprecipitated both from cell extracts and from the medium with antibodies against human placenta Trx. Secretion of newly synthesized Trx was also confirmed by a quantitative radioimmunoassay for human Trx. During 24 h cultivation experiments, treatment with SAC induced a 5-fold increase of the Trx content of normal B lymphocytes as well as in B-CLL cells. Approximately two-thirds of the total amount of the enzyme was released into the medium.