MilliporeSigma
  • Characterization of binding of a specific antagonist, [3H]-SQ 29,548, to soluble thromboxane A2/prostaglandin H2 (TP) receptors in human platelet membranes.
Characterization of binding of a specific antagonist, [3H]-SQ 29,548, to soluble thromboxane A2/prostaglandin H2 (TP) receptors in human platelet membranes.
Life sciences (1992-01-01)
H Monshizadegan, A Hedberg, M L Webb
ABSTRACT
Binding of [3H]-SQ 29,548 was characterized to soluble thromboxane A2/prostaglandin H2 (TP) receptors from human platelet membranes as a means of examining ligand-receptor interactions outside the lipophilic environment of the cell membrane. Kinetic determination revealed a rate of ligand-receptor association of 1.4 x 10(7) +/- 0.2 M-1 x min-1 and a rate of dissociation of 0.5 +/- 0.07 min-1. The resultant equilibrium affinity constant was 36.3 +/- 5.8 nM. Saturation binding analysis revealed a single class of [3H]-SQ 29,548 binding sites with an affinity constant of 39.7 +/- 4.3 nM and a B(max) of 1735.7 +/- 69.1 fmol/mg protein. Specific [3H]-SQ 29,548 binding was inhibited by specific TP receptor antagonists and agonists in a rank order of potency similar to that seen in platelet membranes: SQ 33,961 much greater than SQ 29,548 greater than BM 13,505 greater than or equal to U 46619 greater than BM 13,177. PGD2, PGE2 and PGI2 did not appreciably inhibit the specific binding of [3H]-SQ 29,548. These data indicate that [3H]-SQ 29,548 binding to soluble human platelet TP receptors was specific, saturable, and reversible.