MilliporeSigma
  • Purification and characterization of purine nucleoside phosphorylase and pyrimidine nucleoside phosphorylase from Bacillus stearothermophilus TH 6-2.
Purification and characterization of purine nucleoside phosphorylase and pyrimidine nucleoside phosphorylase from Bacillus stearothermophilus TH 6-2.
Bioscience, biotechnology, and biochemistry (1996-07-01)
T Hamamoto, T Noguchi, Y Midorikawa
ABSTRACT
The purine nucleoside phosphorylase (Pu-NPase) and the pyrimidine nucleoside phosphorylase (Py-NPase) have been purified from Bacillus stearothermophilus TH 6-2. The Pu-NPase is a trimer of 30-kDa subunits and the Py-NPase is a dimer of 46-kDa subunits. The isoelectric points of Pu-NPase and Py-NPase were pH 4.3 and 4.6, respectively. The Pu-NPase could catalyze the phosphorolysis of inosine and guanosine, but not adenosine. the Py-NPase could phosphorolyze both uridine and thymidine.
Sigma-Aldrich
Pyrimidine nucleoside phosphorylase, recombinant, expressed in E. coli, ≥1300 U/mL
Sigma-Aldrich
Pyrimidine Nucleoside Phosphorylase from Bacillus subtilis, recombinant, expressed in E. coli, ≥70 units/mg protein