MilliporeSigma
  • Purification, crystallization, and some properties of creatine amidinohydrolase from Pseudomonas putida.
Purification, crystallization, and some properties of creatine amidinohydrolase from Pseudomonas putida.
Journal of biochemistry (1976-06-01)
T Yoshimoto, I Oka, D Tsuru
PMID8443
ABSTRACT
A method was developed for purification and crystallization of creatinase [creatine amidinohydrolase, EC 3.5.3.3] from Pseudomonas putida var. naraensis C-83. The purified preparation appeared homogeneous on disc electrophoresis and ultracentrifugation and had a molecular weight of 94,000. It was most active at pH 8 and stable between pH 6 and 8 for 24 hr at 37 degrees. SDS-polyacrylamide gel electrophoresis indicated that the native enzyme was made up of two subunit monomers, the molecular weights of which were estimated to be 47,000. Inhibition experiments suggested that a sulfhydryl group is located in or near the active site of the enzyme.
Sigma-Aldrich
Creatinase from microorganisms, lyophilized powder, ≥4 units/mg solid