MilliporeSigma
  • Purification and characterization of choline oxidase from Arthrobacter globiformis.
Purification and characterization of choline oxidase from Arthrobacter globiformis.
Journal of biochemistry (1977-12-01)
S Ikuta, S Imamura, H Misaki, Y Horiuti
PMID599154
ABSTRACT
Choline oxidase was purified from the cells of Arthrobacter globiformis by fractionations with acetone and ammonium sulfate, and column chromatographies on DEAE-cellulose and on Sephadex G-200. The purified enzyme preparation appeared homogeneous on disc gel electrophoresis. The enzyme was a flavoprotein having a molecular weight of approx. 83,000 (gel filtration) or approx. 71,000 (sodium dodecyl sulfate--polyacrylamide disc gel electrophoresis) and an isoelectric point (pI) around pH 4.5. Identification of the reaction products showed that the enzyme catalyzed the following reactions: choline + O2 leads to betaine aldehyde + H2O2, betaine aldehyde + O2 + H2O leads to betaine + H2O2. The enzyme was highly specific for choline and betaine aldehyde (relative reaction velocities: choline, 100%; betaine aldehyde, 46%; N,N-dimethylaminoethanol, 5.2%; triethanolamine, 2.6%; diethanolamine, 0.8%; monoethanolamine, N-methylaminoethanol, methanol, ethanol, propanol, formaldehyde, acetaldehyde, and propionaldehyde, 0%). Its Km values were 1.2 mM for choline and 8.7 mM for betaine aldehyde. The optimum pH for the enzymic reaction was around pH 7.5.
Sigma-Aldrich
Choline Oxidase from Alcaligenes sp., lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Choline Oxidase from Arthrobacter globiformis, lyophilized powder, 8-20 units/mg solid