MilliporeSigma
  • The 5-hydroxytryptamine6 receptor-selective radioligand [3H]Ro 63-0563 labels 5-hydroxytryptamine receptor binding sites in rat and porcine striatum.
The 5-hydroxytryptamine6 receptor-selective radioligand [3H]Ro 63-0563 labels 5-hydroxytryptamine receptor binding sites in rat and porcine striatum.
Molecular pharmacology (1998-09-09)
F G Boess, C Riemer, M Bös, J Bentley, A Bourson, A J Sleight
ABSTRACT
Ro 63-0563 [4-amino-N-(2,6 bis-methylamino-pyridin-4-yl)-benzene sulfonamide] is a high affinity 5-hydroxytryptamine6 (HT6) receptor antagonist with more than 100-fold selectivity for the 5-HT6 receptor compared with 69 other receptors and binding sites. The present study describes the properties of [3H]Ro 63-0563, the first selective 5-HT6 receptor radioligand. Specific binding of [3H]Ro 63-0563 (nonspecific binding defined in the presence of 10 microM methiothepin) to recombinant rat and human 5-HT6 receptors was saturable, rapid, and reversible with equilibrium dissociation constants (Kd) of 6.8 nM and 4.96 nM, respectively. The pharmacological profile of the rat 5-HT6 receptor labeled with [3H]Ro 63-0563 (methiothepin > D-lysergic acid diethylamide > clozapine approximately Ro 63-0563 > lisuride > ergotamine approximately Ro 04-6790 > 5-HT > amitriptyline approximately metergoline approximately mianserin approximately ritanserin > methysergide > mesulergine) was similar to that obtained by using either [3H]D-lysergic acide diethylamide or [3H]5-HT as radioligand. In equilibrium binding studies with rat striatal membranes, [3H]Ro 63-0563 labeled a single binding site with Kd and Bmax values of 11. 7 nM and 175 fmol/mg protein, respectively. In porcine striatal membranes, [3H]Ro 63-0563 also labeled a single binding site with Kd and Bmax values of 8.0 nM and 130 fmol/mg protein, respectively. The affinities of 14 5-HT6 receptor ligands at this binding site were similar to those found for the recombinant rat and human 5-HT6 receptor, which suggested the presence of 5-HT6 receptors in porcine striatum.
Sigma-Aldrich
Methiothepin mesylate salt, ≥98% (HPLC), solid