MilliporeSigma
  • A soluble form of the high affinity IgE receptor, Fc-epsilon-RI, circulates in human serum.
A soluble form of the high affinity IgE receptor, Fc-epsilon-RI, circulates in human serum.
PloS one (2011-05-06)
Eleonora Dehlink, Barbara Platzer, Alexandra H Baker, Jessica Larosa, Michael Pardo, Peter Dwyer, Elizabeth H Yen, Zsolt Szépfalusi, Samuel Nurko, Edda Fiebiger
ABSTRACT
Soluble IgE receptors are potential in vivo modulators of IgE-mediated immune responses and are thus important for our basic understanding of allergic responses. We here characterize a novel soluble version of the IgE-binding alpha-chain of Fc-epsilon-RI (sFcεRI), the high affinity receptor for IgE. sFcεRI immunoprecipitates as a protein of ∼40 kDa and contains an intact IgE-binding site. In human serum, sFcεRI is found as a soluble free IgE receptor as well as a complex with IgE. Using a newly established ELISA, we show that serum sFcεRI levels correlate with serum IgE in patients with elevated IgE. We also show that serum of individuals with normal IgE levels can be found to contain high levels of sFcεRI. After IgE-antigen-mediated crosslinking of surface FcεRI, we detect sFcεRI in the exosome-depleted, soluble fraction of cell culture supernatants. We further show that sFcεRI can block binding of IgE to FcεRI expressed at the cell surface. In summary, we here describe the alpha-chain of FcεRI as a circulating soluble IgE receptor isoform in human serum.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System, peroxidase substrate