MilliporeSigma
  • Phosphoinositide 3-kinase δ regulates membrane fission of Golgi carriers for selective cytokine secretion.
Phosphoinositide 3-kinase δ regulates membrane fission of Golgi carriers for selective cytokine secretion.
The Journal of cell biology (2010-09-15)
Pei Ching Low, Ryo Misaki, Kate Schroder, Amanda C Stanley, Matthew J Sweet, Rohan D Teasdale, Bart Vanhaesebroeck, Frédéric A Meunier, Tomohiko Taguchi, Jennifer L Stow
ABSTRACT
Phosphoinositide 3-kinase (PI3K) p110 isoforms are membrane lipid kinases classically involved in signal transduction. Lipopolysaccharide (LPS)-activated macrophages constitutively and abundantly secrete proinflammatory cytokines including tumor necrosis factor-α (TNF). Loss of function of the p110δ isoform of PI3K using inhibitors, RNA-mediated knockdown, or genetic inactivation in mice abolishes TNF trafficking and secretion, trapping TNF in tubular carriers at the trans-Golgi network (TGN). Kinase-active p110δ localizes to the Golgi complex in LPS-activated macrophages, and TNF is loaded into p230-labeled tubules, which cannot undergo fission when p110δ is inactivated. Similar blocks in fission of these tubules and in TNF secretion result from inhibition of the guanosine triphosphatase dynamin 2. These findings demonstrate a new function for p110δ as part of the membrane fission machinery required at the TGN for the selective trafficking and secretion of cytokines in macrophages.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by gel-filtration chromatography
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)