MilliporeSigma
Emodin, a protein tyrosine kinase inhibitor from Polygonum cuspidatum.
Journal of natural products (1992-05-01)
H Jayasuriya, N M Koonchanok, R L Geahlen, J L McLaughlin, C J Chang
ABSTRACT
Bioassay-directed fractionation of a Chinese medicinal plant, Polygonum cuspidatum (Polygonaceae), has led to the discovery of an anthraquinone, emodin [1], as a strong inhibitor of a protein tyrosine kinase (p56lck) partially purified from bovine thymus. Comparison of the IC50 values of emodin for protein tyrosine kinase inhibitory activity with physcion [2] and emodin-O8-D-glucoside [3], also isolated from the same plant, reveal the importance of the hydroxyl groups at C-6 and C-8 for the observed activity.
Product Description
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm