MilliporeSigma
  • Regulation of PKD by the MAPK p38delta in insulin secretion and glucose homeostasis.
Regulation of PKD by the MAPK p38delta in insulin secretion and glucose homeostasis.
Cell (2009-01-13)
Grzegorz Sumara, Ivan Formentini, Stephan Collins, Izabela Sumara, Renata Windak, Bernd Bodenmiller, Reshma Ramracheya, Dorothée Caille, Huiping Jiang, Kenneth A Platt, Paolo Meda, Rudolf Aebersold, Patrik Rorsman, Romeo Ricci
ABSTRACT
Dysfunction and loss of insulin-producing pancreatic beta cells represent hallmarks of diabetes mellitus. Here, we show that mice lacking the mitogen-activated protein kinase (MAPK) p38delta display improved glucose tolerance due to enhanced insulin secretion from pancreatic beta cells. Deletion of p38delta results in pronounced activation of protein kinase D (PKD), the latter of which we have identified as a pivotal regulator of stimulated insulin exocytosis. p38delta catalyzes an inhibitory phosphorylation of PKD1, thereby attenuating stimulated insulin secretion. In addition, p38delta null mice are protected against high-fat-feeding-induced insulin resistance and oxidative stress-mediated beta cell failure. Inhibition of PKD1 reverses enhanced insulin secretion from p38delta-deficient islets and glucose tolerance in p38delta null mice as well as their susceptibility to oxidative stress. In conclusion, the p38delta-PKD pathway integrates regulation of the insulin secretory capacity and survival of pancreatic beta cells, pointing to a pivotal role for this pathway in the development of overt diabetes mellitus.
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture