MilliporeSigma
  • Structural analysis of human liver glyceraldehyde-3-phosphate dehydrogenase.
Structural analysis of human liver glyceraldehyde-3-phosphate dehydrogenase.
Acta crystallographica. Section D, Biological crystallography (2005-10-22)
S A Ismail, H W Park
ABSTRACT
The crystal structure of human liver glyceraldehyde-3-phosphate dehydrogenase (GAPDH) has been determined. This structure represents the first moderate-resolution (2.5 A) and crystallographically refined (Rfree = 22.9%) human GAPDH structure. The liver GAPDH structure consists of a homotetramer, each subunit of which is bound to a nicotinamide adenine dinucleotide (NAD+) molecule. The GAPDH enzyme has glycolytic and non-glycolytic functions, both of which are of chemotherapeutic interest. The availability of a high-quality human GAPDH structure is a necessity for structure-based drug design. In this study, structural differences between human liver and skeletal muscle GAPDHs are reported in order to understand how these two enzymes might respond to anti-trypanosomatid GAPDH inhibitors.
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein