MilliporeSigma
  • Expression, regulation, and triglyceride hydrolase activity of Adiponutrin family members.
Expression, regulation, and triglyceride hydrolase activity of Adiponutrin family members.
Journal of lipid research (2005-09-10)
Andrew C Lake, Ying Sun, Jian-Liang Li, Jae Eun Kim, Jeremy W Johnson, Dongmei Li, Tracy Revett, Heather H Shih, Wei Liu, Janet E Paulsen, Ruth E Gimeno
ABSTRACT
Adiponutrin and a related protein, adipocyte triglyceride lipase (ATGL; also known as Desnutrin), were recently described as adipocyte-specific proteins with lipid hydrolase activity. Using bioinformatics, we identified three additional Adiponutrin family members (GS2, GS2-Like, and PNPLA1). Here, we report on the expression, regulation, and activity of GS2 and GS2-Like compared with Adiponutrin and Desnutrin/ATGL. GS2-Like is expressed and regulated in a manner similar to Adiponutrin; however, the absolute levels of mRNA are significantly lower than those of Adiponutrin or Desnutrin/ATGL. GS2 transcripts were identified only in humans and are highly expressed in adipose as well as other tissues. All four proteins show lipase activity in vitro, which is dependent on the presence of the active site serine for Adiponutrin, Desnutrin/ATGL, and GS2. Overexpression of Desnutrin/ATGL, GS2, and GS2-Like, but not Adiponutrin, decreases intracellular triglyceride levels. This is consistent with a function for Desnutrin/ATGL, GS2, and GS2-Like in lipolysis, but not for Adiponutrin. Consistent with previously reported data, Desnutrin/ATGL is upregulated by fasting in adipose tissue, whereas Adiponutrin is downregulated. Additionally, Adiponutrin and GS2-Like, but not Desnutrin/ATGL, are strongly induced in the liver of ob/ob mice. Our data support distinct functions for Adiponutrin and Desnutrin/ATGL and raise the possibility that GS2 may contribute significantly to lipolysis in human adipose tissue.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg