MilliporeSigma
Human hepatic lipase mutations and polymorphisms.
Human mutation (1992-01-01)
R A Hegele, L Tu, P W Connelly
ABSTRACT
Human hepatic lipase (HL) is a 477 residue glycoprotein that hydrolyzes triglycerides from plasma lipoproteins. Familial HL deficiency is a rare recessive disorder that is characterized by premature atherosclerosis and abnormal circulating lipoproteins. While studying the HL gene from the world's index family with HL deficiency, we identified four coding sequence variants of HL, one in each of exons 4, 5, 6, and 8. In this report we present the genetic basis for two new HL gene variants, one in each of exons 3 and 5. All six HL DNA variants are single base pair changes. Two variants (at codons 133 and 202) are diallelic DNA polymorphisms that are silent at the amino acid level. One variant (V73M) is an allele that defines an uncommon HL isoprotein. One variant (N193S) has two alleles of approximately equal frequency in the population that specify two common HL isoproteins. Two variants (S267F and T383M) are rare mutations found to date only in HL deficient subjects and their relatives. Of the six HL variants described to date, only S267F and T383M are associated with hyperlipidemia.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg