MilliporeSigma
  • Site-specific chemical modification of a glycoprotein fragment expressed in yeast.
Site-specific chemical modification of a glycoprotein fragment expressed in yeast.
Methods in molecular biology (Clifton, N.J.) (2011-06-16)
Junpeng Xiao, Thomas J Tolbert
ABSTRACT
Site-specific modification of glycoproteins has wide application in both biochemical and biophysical studies. This method describes the conjugation of synthetic molecules to the N-terminus of a glycoprotein fragment, viz., human immunoglobulin G subclass 1 fragment crystallizable (IgG1 Fc), by native chemical ligation. The glycosylated IgG1 Fc is expressed in a glycosylation-deficient yeast strain. The N-terminal cysteine is generated by the endogenous yeast protease Kex2 in the yeast secretory pathway. The N-terminal cysteine is then conjugated with a biotin thioester to produce a biotinylated, glycosylated IgG1 Fc using native chemical ligation.
Sigma-Aldrich
Antifoam 204, aqueous emulsion for bacterial and mammalian systems