MilliporeSigma
  • Trans-4-aminocrotonic acid (TACA), a potent agonist of GABA(A) and GABA(C) receptors, shows a proconvulsant action in the electroconvulsive threshold test in mice.
Trans-4-aminocrotonic acid (TACA), a potent agonist of GABA(A) and GABA(C) receptors, shows a proconvulsant action in the electroconvulsive threshold test in mice.
Pharmacological reports : PR (2005-04-26)
Kinga K Borowicz, Marek Zadrozniak, Stanisław J Czuczwar
ABSTRACT
In the present study, we evaluated TACA (a potent agonist of GABA(A) and GABA(C) receptors) in the electroconvulsive threshold test in mice. Surprisingly, TACA (at 15 and 25 mg/kg) significantly decreased the threshold. The highest ineffective dose of TACA was estimated as 10 mg/kg. The obtained results indicate that the drug enhancing GABAergic transmission may possess proconvulsant properties in the electroconvulsive test. Such effect was completely opposite to our primary assumption and expectance.
Sigma-Aldrich
(E)-4-Amino-2-butenoic acid