MilliporeSigma
  • SB-431542, a small molecule transforming growth factor-beta-receptor antagonist, inhibits human glioma cell line proliferation and motility.
SB-431542, a small molecule transforming growth factor-beta-receptor antagonist, inhibits human glioma cell line proliferation and motility.
Molecular cancer therapeutics (2004-06-24)
Mark D Hjelmeland, Anita B Hjelmeland, Sith Sathornsumetee, Elizabeth D Reese, Michael H Herbstreith, Nicholas J Laping, Henry S Friedman, Darell D Bigner, Xiao-Fan Wang, Jeremy N Rich
ABSTRACT
Transforming growth factor-beta (TGF-beta) is a multifunctional cytokine that promotes malignant glioma invasion, angiogenesis, and immunosuppression. Antisense oligonucleotide suppression of TGF-beta(2) ligand expression has shown promise in preclinical and clinical studies but at least two ligands mediate the effects of TGF-beta in gliomas. Therefore, we examined the effects of SB-431542, a novel, small molecule inhibitor of the type I TGF-beta receptor, on a panel of human malignant glioma cell lines. SB-431542 blocked the phosphorylation and nuclear translocation of the SMADs, intracellular mediators of TGF-beta signaling, with decreased TGF-beta-mediated transcription. Furthermore, SB-431542 inhibited the expression of two critical effectors of TGF-beta-vascular endothelial growth factor and plasminogen activator inhibitor-1. SB-431542 treatment of glioma cultures inhibited proliferation, TGF-beta-mediated morphologic changes, and cellular motility. Together, our results suggest that small molecule inhibitors of TGF-beta receptors may offer a novel therapy for malignant gliomas by reducing cell proliferation, angiogenesis, and motility.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder