MilliporeSigma
  • Identification and characterization of peptide: N-glycanase from Dictyostelium discoideum.
Identification and characterization of peptide: N-glycanase from Dictyostelium discoideum.
BMC biochemistry (2012-06-12)
Anuradha Gosain, Rakhee Lohia, Anju Shrivastava, Shweta Saran
ABSTRACT
Peptide: N- glycanase (PNGase) enzyme cleaves oligosaccharides from the misfolded glycoproteins and prepares them for degradation. This enzyme plays a role in the endoplasmic reticulum associated degradation (ERAD) pathway in yeast and mice but its biological importance and role in multicellular development remain largely unknown. In this study, the PNGase from the cellular slime mold, Dictyostelium discoideum (DdPNGase) was identified based on the presence of a common TG (transglutaminase) core domain and its sequence homology with the known PNGases. The domain architecture and the sequence comparison validated the presence of probable functional domains in DdPNGase. The tertiary structure matched with the mouse PNGase. Here we show that DdPNGase is an essential protein, required for aggregation during multicellular development and a knockout strain of it results in small sized aggregates, all of which did not form fruiting bodies. The in situ hybridization and RT-PCR results show higher level of expression during the aggregate stage. The expression gets restricted to the prestalk region during later developmental stages. DdPNGase is a functional peptide:N-glycanase enzyme possessing deglycosylation activity, but does not possess any significant transamidation activity. We have identified and characterized a novel PNGase from D. discoideum and confirmed its deglycosylation activity. The results emphasize the importance of PNGase in aggregation during multicellular development of this organism.
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit
Sigma-Aldrich
Transglutaminase Assay Kit, sufficient for assays in two 96-well plates
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Sigma-Aldrich
Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) antibody produced in mouse, clone OVA-14, ascites fluid