MilliporeSigma
  • Monitoring and scoring counter-diffusion protein crystallization experiments in capillaries by in situ dynamic light scattering.
Monitoring and scoring counter-diffusion protein crystallization experiments in capillaries by in situ dynamic light scattering.
PloS one (2012-06-08)
Dominik Oberthuer, Emilio Melero-García, Karsten Dierks, Arne Meyer, Christian Betzel, Alfonso Garcia-Caballero, Jose A Gavira
ABSTRACT
In this paper, we demonstrate the feasibility of using in situ Dynamic Light Scattering (DLS) to monitor counter-diffusion crystallization experiments in capillaries. Firstly, we have validated the quality of the DLS signal in thin capillaries, which is comparable to that obtained in standard quartz cuvettes. Then, we have carried out DLS measurements of a counter-diffusion crystallization experiment of glucose isomerase in capillaries of different diameters (0.1, 0.2 and 0.3 mm) in order to follow the temporal evolution of protein supersaturation. Finally, we have compared DLS data with optical recordings of the progression of the crystallization front and with a simulation model of counter-diffusion in 1D.
Sigma-Aldrich
Glucose Isomerase from Streptomyces murinus, ≥350 U/g