MilliporeSigma
  • Cytochemical investigation of the digestive gland of two strombidae species (Strombus gigas and Strombus pugilis) in relation to the nutrition.
Cytochemical investigation of the digestive gland of two strombidae species (Strombus gigas and Strombus pugilis) in relation to the nutrition.
Microscopy research and technique (2012-05-26)
Jean-Marie Volland, Olivier Gros
ABSTRACT
Strombus gigas and Strombus pugilis are threatened species and aquaculture represents a good alternative solution to the fishing. In this study, we highlighted the intracellular digestion process in the digestive gland of two Strombidae species, S. gigas and Strombuspugilis, by the cytochemical characterization of two lysosomal enzymes: acid phosphatase and arylsulfatase. In order to check the efficiency of artificial food digestion, we conducted the characterization on freshly collected, starved and artificially fed individuals of S. pugilis. TEM observations of digestive gland sections from freshly collected individuals of both species revealed the presence of acid phosphatase and arylsulfatase activity mostly located in the apical third of digestive cells. Both enzymes were also detected in artificially fed individuals. In response to the starvation, acid phosphatase is not produced anymore by digestive cells, while arylsulfatase is still present. To our knowledge, this is the first cytochemical validation of intracellular digestion of artificial food in Strombidae. This study highlights the intracellular digestion of artificial food developed for Strombidae aquaculture. Moreover, we have shown that the lysosomal activity could be used as a feed index.
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Sigma-Aldrich
Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL