MilliporeSigma
  • Carbohydrate digestion in Lutzomyia longipalpis' larvae (Diptera - Psychodidae).
Carbohydrate digestion in Lutzomyia longipalpis' larvae (Diptera - Psychodidae).
Journal of insect physiology (2012-07-31)
Vladimir F Vale, Bruno H Moreira, Caroline S Moraes, Marcos H Pereira, Fernando A Genta, Nelder F Gontijo
ABSTRACT
Lutzomyia longipalpis is the principal species of phlebotomine incriminated as vector of Leishmania infantum, the etiological agent of visceral leishmaniasis in the Americas. Despite its importance as vector, almost nothing related to the larval biology, especially about its digestive system has been published. The objective of the present study was to obtain an overview of carbohydrate digestion by the larvae. Taking in account that phlebotomine larvae live in the soil rich in decaying materials and microorganisms we searched principally for enzymes capable to hydrolyze carbohydrates present in this kind of substrate. The principal carbohydrases encountered in the midgut were partially characterized. One of them is a α-amylase present in the anterior midgut. It is probably involved with the digestion of glycogen, the reserve carbohydrate of fungi. Two other especially active enzymes were present in the posterior midgut, a membrane bound α-glucosidase and a membrane bound trehalase. The first, complete the digestion of glycogen and the other probably acts in the digestion of trehalose, a carbohydrate usually encountered in microorganisms undergoing hydric stress. In a screening done with the use of p-nitrophenyl-derived substrates other less active enzymes were also observed in the midgut. A general view of carbohydrate digestion in L. longipalpis was presented. Our results indicate that soil microorganisms appear to be the main source of nutrients for the larvae.
Sigma-Aldrich
Trehalase from porcine kidney, buffered aqueous glycerol solution, ≥1.0 units/mg protein
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000