MilliporeSigma
  • Development of an industrial medium and a novel fed-batch strategy for high-level expression of recombinant β-mananase by Pichia pastoris.
Development of an industrial medium and a novel fed-batch strategy for high-level expression of recombinant β-mananase by Pichia pastoris.
Bioresource technology (2012-06-19)
Jia Zheng, Wei Zhao, Ning Guo, Fulai Lin, Jian Tian, Lishuang Wu, Hongbo Zhou
ABSTRACT
An industrial medium, Corn Steep Liquor Powder Dextrose (CSD medium) was developed for constitutive expression of recombinant β-mananase by Pichia pastoris. The β-mananase activity (513 U/mL) with CSD medium was 1.64- and 2.5-fold higher than with YPD and BSM in shaken flasks. The β-mananase productivity with CSD medium was 61.0 U/mL h, which was 1.7- and 2.5-fold higher than with YPD and BSM in a 5-L fermenter based on a novel fed-batch strategy combining the real-time exponential feed mode with the DO-stat feed mode. The β-mananase activity, dry cell weight and the recombinant enzyme reached up to 5132 U/mL, 110.0 g/L and 4.50 g/L after 50 h cultivation in a 50-L fermenter. The high efficient expression of recombinant β-mananase by P. pastoris indicated that CSD medium and the novel fed-batch strategy have great potential for the production of recombinant β-mananase in industrial fermentation.
Sigma-Aldrich
β-Mannosidase from Helix pomatia, 5-30 units/mL, ammonium sulfate suspension, crude extract