MilliporeSigma
  • Catabolite control protein A controls hydrogen peroxide production and cell death in Streptococcus sanguinis.
Catabolite control protein A controls hydrogen peroxide production and cell death in Streptococcus sanguinis.
Journal of bacteriology (2010-11-03)
Lanyan Zheng, Zhijun Chen, Andreas Itzek, Michael Ashby, Jens Kreth
ABSTRACT
Streptococcus sanguinis is a commensal oral bacterium producing hydrogen peroxide (H₂O₂) that is dependent on pyruvate oxidase (Spx) activity. In addition to its well-known role in bacterial antagonism during interspecies competition, H₂O₂ causes cell death in about 10% of the S. sanguinis population. As a consequence of H₂O₂-induced cell death, largely intact chromosomal DNA is released into the environment. This extracellular DNA (eDNA) contributes to the self-aggregation phenotype under aerobic conditions. To further investigate the regulation of spx gene expression, we assessed the role of catabolite control protein A (CcpA) in spx expression control. We report here that CcpA represses spx expression. An isogenic ΔccpA mutant showed elevated spx expression, increased Spx abundance, and H₂O₂ production, whereas the wild type did not respond with altered spx expression in the presence of glucose and other carbohydrates. Since H₂O₂ is directly involved in the release of eDNA and bacterial cell death, the presented data suggest that CcpA is a central control element in this important developmental process in S. sanguinis.
Sigma-Aldrich
Pyruvate Oxidase from microorganisms, lyophilized powder, ≥1.5 U/mg
Sigma-Aldrich
Pyruvate Oxidase from Aerococcus sp., lyophilized powder, ≥35 units/mg protein (biuret)