MilliporeSigma
  • [The glutathione system in the blood of rats and morphological changes of the pancreas under experimental acute and chronic pancreatitis].
[The glutathione system in the blood of rats and morphological changes of the pancreas under experimental acute and chronic pancreatitis].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2013-03-29)
V A Makarchuk, H O Ushakova, O O Krylova
ABSTRACT
In experiment on laboratory rats the models of acute and chronic pancreatitis were developed to study the changes of lipoperoxidation-antioxidant protection system depending on morphological changes of the pancreas. The acute and chronic pancreatitis is accompanied with intensification of lipoperoxidation and gradual inhibition of antioxidant system due to development of subsequent chronization of the pathological process.