MilliporeSigma
  • Plasmodium falciparum susceptibility to anti-malarial drugs in Dakar, Senegal, in 2010: an ex vivo and drug resistance molecular markers study.
Plasmodium falciparum susceptibility to anti-malarial drugs in Dakar, Senegal, in 2010: an ex vivo and drug resistance molecular markers study.
Malaria journal (2013-03-21)
Bécaye Fall, Aurélie Pascual, Fatoumata D Sarr, Nathalie Wurtz, Vincent Richard, Eric Baret, Yaya Diémé, Sébastien Briolant, Raymond Bercion, Boubacar Wade, Adama Tall, Bruno Pradines
ABSTRACT
In 2006, the Senegalese National Malaria Control Programme recommended artemisinin-based combination therapy (ACT) as the first-line treatment for uncomplicated malaria. Since the introduction of ACT, there have been very few reports on the level of resistance of P. falciparum to anti-malarial drugs. To determine whether parasite susceptibility has been affected by the new anti-malarial policies, an ex vivo susceptibility and drug resistance molecular marker study was conducted on local isolates obtained from the Centre de santé Elizabeth Diouf (Médina, Dakar, Senegal). The prevalence of genetic polymorphisms in genes associated with anti-malarial drug resistance, i.e., pfcrt, pfdhfr, pfdhps and pfmdr1, were evaluated for a panel of 165 isolates collected from patients recruited from 17 August 2010 to 6 January 2011. The malaria isolates were assessed for susceptibility to chloroquine (CQ); quinine (QN); monodesethylamodiaquine (MDAQ), the active metabolite of amodiaquine; mefloquine (MQ); lumefantrine (LMF); dihydroartemisinin (DHA), the active metabolite of artemisinin derivatives; and doxycycline (DOX) using the Plasmodium lactate dehydrogenase (pLDH) ELISA. The prevalence of the in vitro resistant isolates, or isolates with reduced susceptibility, was 62.1% for MQ, 24.2% for CQ, 10.3% for DOX, 11.8% MDAQ, 9.7% for QN, 2.9% for LMF and 0% for DHA. The Pfcrt 76T mutation was identified in 43.6% of the samples. The pfmdr1 86Y, 184F and 1246Y mutations were found in 16.2%, 50.0% and 1.6% of the samples, respectively. The pfdhfr 108N, 51I and 59R mutations were identified in 81.9%, 77.4% and 79.4% of the samples, respectively. The double mutant (108N and 51I) was detected in 75.5% of the isolates, and the triple mutant (108N, 51I and 59R) was detected in 73.6% of the isolates. The pfdhps 437G, 436A and 613S mutations were found in 54.4%, 38.6% and 1.2% of the samples, respectively. There was only one double mutant, 437G and 540E, and one quintuple mutant, pfdhfr 108N, 51I and 59R and pfdhps 437G and 540E. The prevalence of the quadruple mutant (pfdhfr 108N, 51I and 59R and pfdhps 437G) was 36.7%. The results of this study indicate that an intensive surveillance of the in vitro P. falciparum susceptibility to anti-malarial drugs must be conducted in Senegal.
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein