MilliporeSigma
  • A novel benzo[d]imidazole derivate prevents the development of dextran sulfate sodium-induced murine experimental colitis via inhibition of NLRP3 inflammasome.
A novel benzo[d]imidazole derivate prevents the development of dextran sulfate sodium-induced murine experimental colitis via inhibition of NLRP3 inflammasome.
Biochemical pharmacology (2013-03-20)
Wen Liu, Wenjie Guo, Jing Wu, Qiong Luo, Feifei Tao, Yanhong Gu, Yan Shen, Jianxin Li, Renxiang Tan, Qiang Xu, Yang Sun
ABSTRACT
NLRP3 inflammasome has been reported to be associated with various kinds of immunological diseases including colitis. However, there are few drug candidates targeting inflammasomes for the treatment of colitis. In the present study, we aimed at examining the effect of 1-ethyl-5-methyl-2-phenyl-1H-benzo[d]imidazole, a synthetic small molecular compound also named Fc11a-2, for the treatment of dextran sulfate sodium (DSS)-induced experimental colitis in mice via targeting NLRP3 inflammasome. Treatment with Fc11a-2 dose-dependently attenuated the loss of body weight and shortening of colon length induced by DSS. In addition, the disease activity index, histopathologic scores and myeloperoxidase activity were also significantly reduced by Fc11a-2 treatment. Moreover, protein and mRNA levels of DSS-induced proinflammatory cytokines in colon, including TNF-α, IL-1β, IL-18, IL-17A and IFN-γ, were markedly suppressed by Fc11a-2. Furthermore, a decreased CD11c⁺ macrophage infiltration in colons and inactivation of caspase-1 in peritoneal macrophages were detected in Fc11a-2-treated mice. The mechanism of action of Fc11a-2 was related to the inhibition of the cleavage of pro-caspase-1, pro-IL-1β and pro-IL-18 which in turn suppressed the activation of NLRP3 inflammasome. Taken together, our results demonstrate the ability of Fc11a-2 to inhibit NLRP3 inflammasome activation and its potential use in the treatment of inflammatory bowel diseases.
Sigma-Aldrich
Lactoperoxidase from bovine milk, lyophilized, powder, ≥150 U/mg
Sigma-Aldrich
Peroxidase from horseradish, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Peroxidase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Lactoperoxidase from bovine milk, lyophilized powder (essentially salt-free), ≥200 units/mg protein
Sigma-Aldrich
Peroxidase from horseradish, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol), 950-2000 units/mg solid (using ABTS)
Sigma-Aldrich
Peroxidase from horseradish, Type X, ammonium sulfate suspension
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Myeloperoxidase from human leukocytes, lyophilized powder, ≥50 units/mg protein