MilliporeSigma
  • Outcomes of discontinuing rivaroxaban compared with warfarin in patients with nonvalvular atrial fibrillation: analysis from the ROCKET AF trial (Rivaroxaban Once-Daily, Oral, Direct Factor Xa Inhibition Compared With Vitamin K Antagonism for Prevention of Stroke and Embolism Trial in Atrial Fibrillation).
Outcomes of discontinuing rivaroxaban compared with warfarin in patients with nonvalvular atrial fibrillation: analysis from the ROCKET AF trial (Rivaroxaban Once-Daily, Oral, Direct Factor Xa Inhibition Compared With Vitamin K Antagonism for Prevention of Stroke and Embolism Trial in Atrial Fibrillation).
Journal of the American College of Cardiology (2013-02-09)
Manesh R Patel, Anne S Hellkamp, Yuliya Lokhnygina, Jonathan P Piccini, Zhongxin Zhang, Surya Mohanty, Daniel E Singer, Werner Hacke, Günter Breithardt, Jonathan L Halperin, Graeme J Hankey, Richard C Becker, Christopher C Nessel, Scott D Berkowitz, Robert M Califf, Keith A A Fox, Kenneth W Mahaffey
ABSTRACT
The purpose of this study was to understand the possible risk of discontinuation in the context of clinical care. Rivaroxaban is noninferior to warfarin for preventing stroke in atrial fibrillation patients. Concerns exist regarding possible increased risk of stroke and non-central nervous system (CNS) thromboembolic events early after discontinuation of rivaroxaban. We undertook a post-hoc analysis of data from the ROCKET AF (Rivaroxaban Once-Daily, Oral, Direct Factor Xa Inhibition Compared With Vitamin K Antagonism for Prevention of Stroke and Embolism Trial in Atrial Fibrillation, n = 14,624) for stroke or non-CNS embolism within 30 days after temporary interruptions of 3 days or more, early permanent study drug discontinuation, and end-of-study transition to open-label therapy. Stroke and non-CNS embolism occurred at similar rates after temporary interruptions (rivaroxaban: n = 9, warfarin: n = 8, 6.20 vs. 5.05/100 patient-years, hazard ratio [HR]: 1.28, 95% confidence interval [CI]: 0.49 to 3.31, p = 0.62) and after early permanent discontinuation (rivaroxaban: n = 42, warfarin: n = 36, 25.60 vs. 23.28/100 patient-years, HR: 1.10, 95% CI: 0.71 to 1.72, p = 0.66). Patients transitioning to open-label therapy at the end of the study had more strokes with rivaroxaban (n = 22) versus warfarin (n = 6, 6.42 vs. 1.73/100 patient-years, HR: 3.72, 95% CI: 1.51 to 9.16, p = 0.0044) and took longer to reach a therapeutic international normalized ratio with rivaroxaban versus warfarin. All thrombotic events within 30 days of any study drug cessation (including stroke, non-CNS embolism, myocardial infarction, and vascular death) were similar between groups (HR: 1.02, 95% CI: 0.83 to 1.26, p = 0.85). In atrial fibrillation patients who temporarily or permanently discontinued anticoagulation, the risk of stroke or non-CNS embolism was similar with rivaroxaban or warfarin. An increased risk of stroke and non-CNS embolism was observed in rivaroxaban-treated patients compared with warfarin-treated patients after the end of the study, underscoring the importance of therapeutic anticoagulation coverage during such a transition.
Sigma-Aldrich
Factor X Activated (Xa) from bovine plasma, aqueous glycerol solution