MilliporeSigma
  • An extra peptide within the catalytic module of a β-agarase affects the agarose degradation pattern.
An extra peptide within the catalytic module of a β-agarase affects the agarose degradation pattern.
The Journal of biological chemistry (2013-02-05)
Wen-Jun Han, Jing-Yan Gu, Hui-Hui Liu, Fu-Chuan Li, Zhi-Hong Wu, Yue-Zhong Li
ABSTRACT
Agarase hydrolyzes agarose into a series of oligosaccharides with repeating disaccharide units. The glycoside hydrolase (GH) module of agarase is known to be responsible for its catalytic activity. However, variations in the composition of the GH module and its effects on enzymatic functions have been minimally elucidated. The agaG4 gene, cloned from the genome of the agarolytic Flammeovirga strain MY04, encodes a 503-amino acid protein, AgaG4. Compared with elucidated agarases, AgaG4 contains an extra peptide (Asn(246)-Gly(302)) within its GH module. Heterologously expressed AgaG4 (recombinant AgaG4; rAgaG4) was determined to be an endo-type β-agarase. The protein degraded agarose into neoagarotetraose and neoagarohexaose at a final molar ratio of 1.5:1. Neoagarooctaose was the smallest substrate for rAgaG4, whereas neoagarotetraose was the minimal degradation product. Removing the extra fragment from the GH module led to the inability of the mutant (rAgaG4-T57) to degrade neoagarooctaose, and the final degradation products of agarose by the truncated protein were neoagarotetraose, neoagarohexaose, and neoagarooctaose at a final molar ratio of 2.7:2.8:1. The optimal temperature for agarose degradation also decreased to 40 °C for this mutant. Bioinformatic analysis suggested that tyrosine 276 within the extra fragment was a candidate active site residue for the enzymatic activity. Site-swapping experiments of Tyr(276) to 19 various other amino acids demonstrated that the characteristics of this residue were crucial for the AgaG4 degradation of agarose and the cleavage pattern of substrate.
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarase from Pseudomonas atlantica, lyophilized powder, ≥5,000 units/mg protein (Lowry)
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter