MilliporeSigma
  • Mechanism of the glycosylation step catalyzed by human α-galactosidase: a QM/MM metadynamics study.
Mechanism of the glycosylation step catalyzed by human α-galactosidase: a QM/MM metadynamics study.
The journal of physical chemistry. B (2012-12-20)
Xiao-Liang Pan, Wei Liu, Jing-Yao Liu
ABSTRACT
The enzyme α-galactosidase (α-GAL), a member of glycoside hydrolase family 27, catalyzes the removal of a nonreducing terminal α-galactose residue from polysaccharides, glycolipids, and glycopeptides. α-GAL is believed to have the double displacement retaining reaction mechanism. In this work, the glycosylation step catalyzed by human α-GAL was computationally simulated with quantum mechanics/molecular mechanics metadynamics. Our simulations show that the overall catalytic mechanism follows a D(N)*A(N)-like mechanism, and the transition state has a oxocarbenium ion like character with a partially formed double bond between the ring oxygen and C5' carbon atoms. In addition, the galactosyl ring of the substrate follows a conformational itinerary of (4)C(1) → [E(3)/(4)H(3)](++) → (1)S(3) along the reaction coordinate.
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
α-Galactosidase from green coffee beans, ammonium sulfate suspension, ≥9 units/mg protein