MilliporeSigma
Working memory is differentially affected by stress in men and women.
Behavioural brain research (2012-12-15)
Daniela Schoofs, Stephan Pabst, Matthias Brand, Oliver T Wolf
ABSTRACT
Stress has been shown to influence working memory. However, sex differences and the potential impact of stimulus emotionality have not received much attention. In a first experiment the effects of stress on a neutral working memory (WM) paradigm were tested in male and female participants (Experiment 1). Experiment 2 employed the same paradigm but used emotional stimuli. For this purpose, healthy participants were exposed either to a stressful (Trierer Social Stress Test (TSST)) or to a non-stressful control condition. Subsequently, WM performance in an n-back task was assessed. In Experiment 1, single digits were used as stimuli, while in Experiment 2 neutral and negative pictures were additionally employed. Salivary cortisol and Alpha-Amylase (sAA) were measured before and three times after the treatment as a marker of hypothalamus-pituitary-adrenal (HPA) axis- and sympathetic nervous system (SNS) activity. In both experiments, stress caused a substantial cortisol and sAA increase. For WM performance (response time) a stress by sex interaction was apparent. Stress enhanced performance in men, while impairing it in women. In both experiments stress had no effect on response accuracy. No modulating effect of the emotional quality of stimuli on n-back performance was observed (study 2). The results indicate that the effect of acute stress on n-back performance differs between the sexes. In contrast to long-term memory, the influence of stress on WM appears not to be modulated by the emotionality of the employed stimuli if stimuli are potential targets as it is the case in the n-back task.
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid