MilliporeSigma
  • Enzymatic hydrolysis of microcrystalline cellulose and pretreated wheat straw: a detailed comparison using convenient kinetic analysis.
Enzymatic hydrolysis of microcrystalline cellulose and pretreated wheat straw: a detailed comparison using convenient kinetic analysis.
Bioresource technology (2012-12-12)
Mareike Monschein, Christoph Reisinger, Bernd Nidetzky
ABSTRACT
Marked slow-down of soluble sugar production at low degree of substrate conversion limits the space-time yield of enzymatic hydrolysis of ligno-cellulosic materials. A simple set of kinetic descriptors was developed to compare reducing sugar release from pure crystalline cellulose (Avicel) and pretreated wheat straw by Trichoderma reesei cellulase at 50 °C. The focus was on the rate-retarding effect of maximum hydrolysis rate at reaction start (r(max)), limiting hydrolysis rate (r(lim)) at extended reaction time (24h), and substrate conversion, marking the transition between the r(max) and r(lim) kinetic regimes (C(trans)). At apparent saturation of substrate (12.2g cellulose/L) with enzyme, r(max) for pretreated wheat straw (~9.6g/L/h) surpassed that for Avicel by about 1.7-fold whereas their r(lim) were almost identical (~0.15 g/L/h). C(trans) roughly doubled as enzyme/substrate loading was increased from 3.8 to 75FPU/g, suggesting C(trans) to be a complex manifestation of cellulase-cellulose interaction, not an intrinsic substrate property. A low-temperature adsorption step preceding hydrolysis at 50 °C resulted in enhanced cellulase binding at reaction start without increasing r(max). C(trans) was higher for pretreated wheat straw (~30%) than for Avicel (~20%) under these conditions.
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg