MilliporeSigma
  • Glycosylated copper(II) ionophores as prodrugs for β-glucosidase activation in targeted cancer therapy.
Glycosylated copper(II) ionophores as prodrugs for β-glucosidase activation in targeted cancer therapy.
Dalton transactions (Cambridge, England : 2003) (2012-11-24)
Valentina Oliveri, Maurizio Viale, Giulia Caron, Cinzia Aiello, Rosaria Gangemi, Graziella Vecchio
ABSTRACT
8-Hydroxyquinoline derivatives are metal-binding compounds that have recently attracted interest as therapeutic agents for cancer therapy. In this scenario, we designed and synthesized three new glucoconjugates, 5,7-dichloro-8-quinolinyl-β-D-glucopyranoside, 5-chloro-8-quinolinyl-β-D-glucopyranoside and 2-methyl-8-quinolinyl-β-D-glucopyranoside and investigated their biological properties in comparison to the parent 8-hydroxyquinoline derivatives in the presence of Cu(2+). In vitro data show that 2 out of 3 glycosylated compounds possess a pharmacologically-relevant antiproliferative activity against tumor cells, similar to that of their parent compounds; this activity is associated with a relevant triggering of apoptosis. The pharmacological profile of the glucoconjugates depends on the cellular enzymatic β-glucosidase activity, as demonstrated by the inhibition of antiproliferative activity in the presence of the 2,5-dideoxy-2,5-imino-D-mannitol.
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, ≥2 units/mg solid
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized, powder, ≥4 U/mg
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, 10-50 units/mg solid