MilliporeSigma
  • Synthesis of novel 12-aryl-8,9,10,12-tetrahydrobenzo[a]xanthene-11-thiones and evaluation of their biocidal effects.
Synthesis of novel 12-aryl-8,9,10,12-tetrahydrobenzo[a]xanthene-11-thiones and evaluation of their biocidal effects.
European journal of medicinal chemistry (2012-11-17)
Jitender M Khurana, Devanshi Magoo, Komal Aggarwal, Nisha Aggarwal, Rajesh Kumar, Chitra Srivastava
ABSTRACT
Novel 12-aryl-8,9,10,12-tetrahydrobenzo[a]xanthene-11-thiones have been synthesized in high yields by treatment of the corresponding oxo analogs with Lawesson's reagent. The structure has been confirmed by X-ray analysis. The compounds were tested for in vitro antifungal activity against Rhizoctonia bataticola, Sclerotium rolfsii, Fusarium oxysporum and Alternaria porii. The compounds exhibited moderate to good activity against all pathogens. Insecticidal activity of these compounds against Spodoptera litura was observed to be comparable to commercial pyrethroid insecticide, cypermethrin. The urease inhibitory activity has also been studied.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg