MilliporeSigma
  • Purification and biochemical characterisation of a glucose-6-phosphate dehydrogenase from the psychrophilic green alga Koliella antarctica.
Purification and biochemical characterisation of a glucose-6-phosphate dehydrogenase from the psychrophilic green alga Koliella antarctica.
Extremophiles : life under extreme conditions (2012-11-03)
Myriam Ferrara, Gea Guerriero, Manuela Cardi, Sergio Esposito
ABSTRACT
Psychrophilic organisms have evolved a number of modifications of cellular structures to survive in the cold environment; among them it is worth noting an increased efficiency of enzymes at lower temperatures. Glucose-6-phosphate dehydrogenase (G6PDH; EC 1.1.1.49) was purified and characterised from the psychrophilic green alga Koliella antarctica (Trebouxiophyceae, Chlorophyta) from the Ross Sea (Antarctica). It was possible to isolate a single G6PDH using biochemical strategies; its maximum activity was measured at 35 °C, and the enzyme showed an E (a) of 39.6 kJ mol(-1). This protein reacted with antibodies raised against higher plants plastidic isoforms. KaG6PDH showed peculiar kinetic properties, with a K (iNADPH) value lower than [Formula: see text]. Notably, catalytic activity was inactivated in vitro by DTT and chloroplastic thioredoxin f. These biochemical properties of G6PDH are discussed with respect to higher plant G6PDHs and the adaptation of K. antarctica to polar low-temperature environment.
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret