MilliporeSigma
  • An antigenic domain within a catalytically active Leishmania infantum nucleoside triphosphate diphosphohydrolase (NTPDase 1) is a target of inhibitory antibodies.
An antigenic domain within a catalytically active Leishmania infantum nucleoside triphosphate diphosphohydrolase (NTPDase 1) is a target of inhibitory antibodies.
Parasitology international (2012-09-22)
Ana Carolina Ribeiro Gomes Maia, Gabriane Nascimento Porcino, Michelle de Lima Detoni, Nayara Braga Emídio, Danielle Gomes Marconato, Priscila Faria-Pinto, Melissa Regina Fessel, Alexandre Barbosa Reis, Luiz Juliano, Maria Aparecida Juliano, Marcos José Marques, Eveline Gomes Vasconcelos
ABSTRACT
We identified a shared B domain within nucleoside triphosphate diphosphohydrolases (NTPDases) of plants and parasites. Now, an NTPDase activity not affected by inhibitors of adenylate kinase and ATPases was detected in Leishmania infantum promastigotes. By non-denaturing gel electrophoresis of detergent-homogenized promastigote preparation, an active band hydrolyzing nucleosides di- and triphosphate was visualized and, following SDS-PAGE and silver staining was identified as a single polypeptide of 50kDa. By Western blots, it was recognized by immune sera raised against potato apyrase (SA), r-pot B domain (SB), a recombinant polypeptide derived from the potato apyrase, and LbB1LJ (SC) or LbB2LJ (SD), synthetic peptides derived from the Leishmania NTPDase 1, and by serum samples from dogs with visceral leishmaniasis, identifying the antigenic L. infantum NTPDase 1 and, also, its conserved B domain (r83-122). By immunoprecipitation assays and Western blots, immune sera SA and SB identified the catalytically active NTPDase 1 in promastigote preparation. In addition, the immune sera SB (44%) and SC or SD (87-99%) inhibited its activity, suggesting a direct effect on the B domain. By ELISA, 37%, 45% or 50% of 38 infected dogs were seropositive for r-pot B domain, LbB1LJ and LbB2LJ, respectively, confirming the B domain antigenicity.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder