MilliporeSigma
  • Catalytic mechanism of porphobilinogen synthase: the chemical step revisited by QM/MM calculations.
Catalytic mechanism of porphobilinogen synthase: the chemical step revisited by QM/MM calculations.
The journal of physical chemistry. B (2012-09-15)
Bo-Xue Tian, Edvin Erdtman, Leif A Eriksson
ABSTRACT
Porphobilinogen synthase (PBGS) catalyzes the asymmetric condensation and cyclization of two 5-aminolevulinic acid (5-ALA) substrate molecules to give porphobilinogen (PBG). The chemical step of PBGS is herein revisited using QM/MM (ONIOM) calculations. Two different protonation states and several different mechanisms are considered. Previous mechanisms based on DFT-only calculations are shown unlikely to occur. According to these new calculations, the deprotonation step rather than ring closure is rate-limiting. Both the C-C bond formation first mechanism and the C-N bond formation first mechanism are possible, depending on how the A-site ALA binds to the enzyme. We furthermore propose that future work should focus on the substrate binding step rather than the enzymatic mechanism.