MilliporeSigma
  • Adsorption characteristics of fungal family 1 cellulose-binding domain from Trichoderma reesei cellobiohydrolase I on crystalline cellulose: negative cooperative adsorption via a steric exclusion effect.
Adsorption characteristics of fungal family 1 cellulose-binding domain from Trichoderma reesei cellobiohydrolase I on crystalline cellulose: negative cooperative adsorption via a steric exclusion effect.
Langmuir : the ACS journal of surfaces and colloids (2012-09-07)
Naohisa Sugimoto, Kiyohiko Igarashi, Masahisa Wada, Masahiro Samejima
ABSTRACT
Cellobiohydrolases (CBHs) hydrolyzing crystalline cellulose share a two-domain structure of catalytic domain (CD) and cellulose-binding domain (CBD). To focus on the binding characteristics of CBD, we analyzed the adsorption of fusion protein of fungal family 1 CBD from Trichoderma reesei CBH I and red-fluorescent protein on crystalline and amorphous celluloses. Binding data were better fitted by Hill's model with negative cooperativity than by other adsorption models, suggesting the occurrence of a steric exclusion effect among the fusion molecules on the cellulose surfaces. The degree of negative cooperativity depended on the nature of the cellulose. The significance of this phenomenon for catalysis by intact CBHI is discussed.
Sigma-Aldrich
Cellobiohydrolase I from Hypocrea jecorina, 100 U/mg, recombinant, expressed in corn
Pricing and availability is not currently available.