MilliporeSigma
  • Effects of milk casein derived tripeptides on endothelial enzymes in vitro; a study with synthetic tripeptides.
Effects of milk casein derived tripeptides on endothelial enzymes in vitro; a study with synthetic tripeptides.
Arzneimittel-Forschung (2012-08-25)
A Siltari, A S Kivimäki, P I Ehlers, R Korpela, H Vapaatalo
ABSTRACT
In the fermentation of milk by certain lactic acid bacteria, casein is degraded into bioactive tripeptides shown to lower blood pressure in experimental animal models and in mildly hypertensive humans. This effect is suggested to result mainly in inhibition of angiotensin converting enzyme 1 (ACE-1).Due to the complexity of renin-angiotensin system (RAS), several other enzymes than ACE-1 can participate in the production of vasoactive components. Therefore, in the present study we investigated effects of tripeptides isoleucine-proline-proline (IPP), valine-proline-proline (VPP) and leucine-proline-proline (LPP) on some endothelial enzymes that are important in RAS or otherwise have a role in the endothelial function. The enzymes investigated were renin, chymase, neutral endopeptidase (NEP), prolyl oligopeptidase (POP), cathepsin G, endothelin converting enzyme 1 (ECE-1), and cyclooxygenase 1 and 2 (COX -1 and COX-2).The tripeptides inhibited prolyl oligopeptidase (POP) dose-dependently. IPP was the most potent inhibitor (IC50 486±95 µM). Contrary, cathepsin G was activated by IPP, VPP and LPP as well as the amino acids proline and isoleucine. The other investigated enzymes were not affected. Inhibition of POP and activation of cathepsin G do not explain the blood pressure lowering effects of the tripeptides. Thus the inhibition of ACE-1 remains the most plausible mechanism of the antihypertensive effects of the tripeptides.
Sigma-Aldrich
Chymase human, recombinant, expressed in Pichia pastoris
Sigma-Aldrich
Cathepsin G from human leukocytes, lyophilized powder, ≥60 units/mg protein (Bradford)