MilliporeSigma
  • Preliminary crystallographic analysis of glyceraldehyde-3-phosphate dehydrogenase 3 from Saccharomyces cerevisiae.
Preliminary crystallographic analysis of glyceraldehyde-3-phosphate dehydrogenase 3 from Saccharomyces cerevisiae.
Acta crystallographica. Section F, Structural biology and crystallization communications (2012-08-08)
Qiao Liu, Hong Wang, Huihui Liu, Maikun Teng, Xu Li
ABSTRACT
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is an important enzyme in the glycolytic pathway. In addition to its conventional metabolic role, GAPDH has been identified to possess diverse cellular functions. In this study, glyceraldehyde-3-phosphate dehydrogenase 3, the third isoform of GAPDH from Saccharomyces cerevisiae, was cloned, expressed, purified and crystallized. The crystals belonged to space group I4(1)22, with unit-cell parameters a = b = 116.13, c = 119.21 Å. X-ray diffraction data were collected to a resolution of 2.6 Å. The structure was solved by molecular replacement and refinement is in progress.
Product Description
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from human erythrocytes, lyophilized powder, 50-150 units/mg protein