MilliporeSigma
  • Expression, purification, crystallization and preliminary X-ray analysis of 4-hydroxy-3-methyl-2-keto-pentanoate aldolase (asHPAL) from Arthrobacter simplex strain AKU 626.
Expression, purification, crystallization and preliminary X-ray analysis of 4-hydroxy-3-methyl-2-keto-pentanoate aldolase (asHPAL) from Arthrobacter simplex strain AKU 626.
Acta crystallographica. Section F, Structural biology and crystallization communications (2012-08-08)
Linjun Guo, Masahiko Okai, Tomoko Mase, Fabiana Lica Imai, Takuya Miyakawa, Koji Nagata, Hiroyuki Yamanaka, Hidemi Fujii, Makoto Hibi, Jun Ogawa, Masaru Tanokura
ABSTRACT
4-Hydroxy-3-methyl-2-keto-pentanoate aldolase (asHPAL), an enzyme used in the synthesis of (2S,3R,4S)-4-hydroxyisoleucine, was crystallized in the absence and the presence of 2-ketobutyrate as one of its substrates by the sitting-drop vapour-diffusion method using PEG 400 as a precipitant. Crystals of asHPAL grown without and with 2-ketobutyrate diffracted to 1.60 and 1.55 Å resolution and belonged to space group C2, with unit-cell parameters a = 116.8, b = 88.2, c = 85.3 Å, β = 122.3° and a = 116.2, b = 88.1, c = 85.0 Å, β = 122.3°, respectively.
Sigma-Aldrich
Aldolase from rabbit muscle, lyophilized powder, ≥8.0 units/mg protein
Sigma-Aldrich
Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein